missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Monobasic, Honeywell™

Product Code. 15661580
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

for HPLC,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7722-76-1
Quantity 250 g
Molecular Formula H6NO4P
Linear Formula NH4H2PO4
MDL Number MFCD00003396
UN Number NONH for all modes of transport
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
Solubility Information Soluble in water 0.1g/mL at 25°C
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES N.OP(O)(O)=O
IUPAC Name azanium;dihydrogen phosphate
Molecular Weight (g/mol) 115.03
PubChem CID 24402
ChEBI CHEBI:62982
Formula Weight 115.03g/mol
Percent Purity ≥99% (T)
Chemical Name or Material Ammonium Phosphate Monobasic
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'