missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulfate, 99%, pure, Thermo Scientific Chemicals
106.53 € - 393.96 €
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12085390
|
Thermo Scientific Acros
446450050 |
5 kg | Plastic Bottle |
106.53 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12095390
|
Thermo Scientific Acros
446450250 |
25 kg | Fibre drum |
393.96 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 |
Specifications
280.0°C | |
99% | |
(NH4)2SO4 | |
15,552 | |
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
99% | |
Ammonium sulfate |
7783-20-2 | |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
6097028 | |
132.14 | |
Pure |
Safety and Handling
missing translation for 'einecsNumber' : 231-984-1
Product Identifier
- Ammonium sulfate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title