Learn More
Ammonium sulfate, ultrapure, 99+%, Thermo Scientific Chemicals
25.02 € - 78.89 €
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
IUPAC Name | sulfuric acid diamine |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426379
|
Thermo Scientific Alfa Aesar
J64419.22 |
100 g |
25.02 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15436379
|
Thermo Scientific Alfa Aesar
J64419.36 |
500 g |
35.07 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446379
|
Thermo Scientific Alfa Aesar
J64419.A3 |
2 kg |
78.89 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ammonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAmmonium sulfate is a widely used reagent in molecular biology and chromatography. It is useful in the isolation and purification of enzymes and proteins. It is used for the precipitation or fractionation of proteins or for purification of antibodies. It is also useful for crystallographic analysis of nucleic acids and proteins. Ammonium sulfate is also widely used in HPLC of proteins, such as in hydrophobic interaction chromatography. Ammonium sulfate is used in long PCR buffer, in PCR lysis solution, and in secondstrand cDNA synthesis. Also it is used for the HPLC analysis of phospholipids.
Solubility
Soluble freely in water, Insoluble in alcohol and acetone.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
sulfuric acid diamine |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 | |
N.N.OS(O)(=O)=O |
Specifications
∽280°C (decomposition) | |
White | |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
sulfuric acid diamine | |
6097028 | |
132.14 | |
1.769 | |
Ultrapure |
7783-20-2 | |
Crystalline | |
(NH4)2SO4 | |
14,555 | |
Soluble freely in water,Insoluble in alcohol and acetone. | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
≥99% | |
Ammonium sulfate |
Safety and Handling
missing translation for 'einecsNumber' : 231-984-1
missing translation for 'rtecsNumber' : BS4500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only