Learn More
Ammonium tetrathiomolybdate, 99.95%, (trace metal basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 300.0°C |
Color | White |
Physical Form | Crystals or Powder |
Quantity | 10 g |
Assay Percent Range | 99.95% |
Linear Formula | (NH4)2MoS4 |
Merck Index | 15,557 |
IUPAC Name | diazanium;bis(sulfanylidene)molybdenum;sulfanide |
Formula Weight | 260.28 |
Percent Purity | 99.95% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0441052 | Certificate of Analysis | 11/06/2024 | 389530500,389532500,389530010,389530100 |
A0432471 | Certificate of Analysis | 25/08/2023 | 389530500,389532500,389530010,389530100 |
A0407593 | Certificate of Analysis | 13/12/2022 | 389530500,389532500,389530010,389530100 |
A0414363 | Certificate of Analysis | 04/05/2022 | 389530500,389532500,389530010,389530100 |
A0441052 | Certificate of Analysis | 11/04/2022 |
Safety and Handling

- Ammonium tetrathiomolybdate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.