missing translation for 'onlineSavingsMsg'
Learn More

Ammonium tetrathiotungstate, Thermo Scientific Chemicals

84.82 € - 338.69 €

Chemical Identifiers

CAS 13862-78-7
Molecular Formula H8N2S4W-2
Molecular Weight (g/mol) 348.158
MDL Number MFCD00061410
InChI Key CCFCMIWYHMKPAO-UHFFFAOYSA-L
Synonym ammonium tetrathiotungstate, ammonium tetrathiotungstate vi, h2s4w.2nh3, disulfanylidenetungstendithiol diamine, ammonium tetrathiotungstate trace metals basis
PubChem CID 53393497
IUPAC Name azane;bis(sulfanylidene)tungsten;sulfanide
SMILES N.N.[SH-].[SH-].S=[W]=S
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11388027
View Documents
Thermo Scientific Alfa Aesar
022373.04
2 g
84.82 €
2g
Please to purchase this item. Need a web account? Register with us today!
11378027
View Documents
Thermo Scientific Alfa Aesar
022373.09
10 g
338.69 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ammonium tetrathiotungstate is used to classic building block for transition metal sulfide cluster compounds was recently used in a low-temperature, solid state synthesis of a new series of heterometallic sulfide clusters stabilized with dppm [bis(diphenylphosphino)methane] ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13862-78-7
348.158
CCFCMIWYHMKPAO-UHFFFAOYSA-L
53393497
N.N.[SH-].[SH-].S=[W]=S
H8N2S4W-2
MFCD00061410
ammonium tetrathiotungstate, ammonium tetrathiotungstate vi, h2s4w.2nh3, disulfanylidenetungstendithiol diamine, ammonium tetrathiotungstate trace metals basis
azane;bis(sulfanylidene)tungsten;sulfanide

Specifications

13862-78-7
Chunks and/or Powder
MFCD00061410
Insoluble in water.
N.N.[SH-].[SH-].S=[W]=S
348.158
348.07
Hygroscopic
Yellow
H8N2S4W-2
ammonium tetrathiotungstate, ammonium tetrathiotungstate vi, h2s4w.2nh3, disulfanylidenetungstendithiol diamine, ammonium tetrathiotungstate trace metals basis
CCFCMIWYHMKPAO-UHFFFAOYSA-L
azane;bis(sulfanylidene)tungsten;sulfanide
53393497
Like rotten eggs
Ammonium tetrathiotungstate
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only