missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate concentrate, For 1L standard solution, 0.1 M NH4SCN (0.1N), Honeywell Fluka™

Product Code. 15615190
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.

for 1L standard solution, 0.1 M NH4SCN (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Quantity 1 Pc.
Linear Formula NH4SCN
UN Number UN3082
Beilstein 3595135
Formula Weight 76.12g/mol
Packaging Ampoule
Chemical Name or Material Ammonium thiocyanate concentrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'