Learn More
Ammonium thioglycolate, 70 wt.% solution in water, Thermo Scientific Chemicals
Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5
27.03 € - 61.41 €
Specifications
Chemical Name or Material | Ammonium thioglycolate |
---|---|
Color | Colorless |
Name Note | 70 wt.% solution in water |
CAS | 5421-46-5, 7732-18-5 |
Boiling Point | 115.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10268713
|
Thermo Scientific Acros
384682500 |
250 mL | Glass bottle |
27.03 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10521334
|
Thermo Scientific Acros
384680010 |
1 L | Glass bottle |
61.41 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium thioglycolate | |
70 wt.% solution in water | |
115.0°C | |
C2H7NO2S | |
1.2200g/mL | |
Solubility in water: completely soluble. | |
C(C(=O)[O-])S.[NH4+] | |
109.143 | |
109.15 | |
ammonium: positive, mercaptane: positive | |
1.22 | |
Solution |
Colorless | |
5421-46-5, 7732-18-5 | |
-20.0°C | |
MFCD00137451 | |
ammonium thioglycolate, ammonium mercaptoacetate, acetic acid, mercapto-, monoammonium salt, thiofaco a-50, ammonium thioglycollate, thioglycolic acid ammonium salt, usaf mo-2, unii-6p9e788vzz, thioglycollic acid, ammonium salt, ammonium 2-mercaptoacetate | |
ZZTCCAPMZLDHFM-UHFFFAOYSA-N | |
azanium;2-sulfanylacetate | |
21534 | |
68.0 to 72.0% | |
Glass bottle | |
5.8 to 6.4 |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May be corrosive to metals.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Ammonium thioglycolate, 70 wt.% solution in water
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Substances and mixtures corrosive to metals Category 1
- Skin sensitiser Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Ammonium thioglycolate
RUO – Research Use Only