missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium thiosulfate, 96%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
Melting Point | 150.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 10 kg |
Assay Percent Range | 95% min. (Iodimetry) |
Linear Formula | (NH4)2S2O3 |
Merck Index | 15, 559 |
Solubility Information | Solubility in water: 63.1% (20°C) - 71.0% (70°C). Other solubilities: insoluble in alcohol and ether |
Formula Weight | 148.21 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- Ammonium thiosulfate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'