missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Trifluoroacetate, ≥99.0% (NT), Honeywell™ Fluka™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 123°C to 125°C |
CAS | 3336-58-1 |
Color | Colorless |
Physical Form | Solid |
Quantity | 1 g |
Molecular Formula | C2H5F3NO2 |
Linear Formula | CF3CO2NH4 |
MDL Number | MFCD00012615,MFCD03095537 |
UN Number | NONH for all modes of transport |
Synonym | ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'