missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMOT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16870-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes p130 only.
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predomitly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
AMOT | |
Polyclonal | |
Unconjugated | |
AMOT | |
AMOT, angiomotin, KIAA1071 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
154796, 27494, 300289 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4VCS5, Q8VHG2 | |
AMOT | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |