missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ampicillin Sodium Salt (Crystalline Powder), Fisher BioReagents™
35.05 EUR valid until 2024-12-31
Use promo code "21971" to get your promotional price.
Use promo code "21971" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Poly Bottle
Description
For selection for ampicillin resistance in mutated and transformed cells
For selection for ampicillin resistance in mutated and transformed cells. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Type | Ampicillin Sodium Salt |
Physical Form | Solid |
pH | 8.0 to 10.0 |
Melting Point | 215°C |
Merck Index | 15, 583 |
Packaging | Poly Bottle |
Potency | Inclusive between 845μg/mg |
Formula Weight | 371.39 |
Water | 2% Max. |
Color | White |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Ampicillin Sodium Salt
- Danger
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'