missing translation for 'onlineSavingsMsg'
Learn More

Ampicillin Sodium Salt, Ultrapure, Thermo Scientific™

Product Code. 15845088
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-52-3
Molecular Formula C16H21N3NaO4S
Molecular Weight (g/mol) 374.411
MDL Number MFCD00064313
InChI Key BSFVNXCYXDYHOD-ZQDFAFASSA-N
Synonym ampicillin sodium
PubChem CID 131673879
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium
SMILES [HH].CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C.[Na]

Specifications

Melting Point 215°C
Physical Form Powder
Quantity 5 g
Beilstein 4119211
Formula Weight 371.39
Grade Ultrapure
Packaging Plastic bottle
Chemical Name or Material Ampicillin Sodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'