missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ DNA Polymerase with Buffer II and MgCl2

Product Code. p-4926872 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1,000 units
25 x 1,000 units
250 Units
3,000 units
5,000 units
Unit Size:
250 units
25000 units
3000 units
5000 units
Each
This item is not returnable. View return policy

Product Code. 10348092

Brand: Applied Biosystems™ N8080241

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

AmpliTaq Gold DNA Polymerase is a chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation.

AmpliTaq Gold DNA Polymerase with Buffer II and MgCl2 is a chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation. The modified enzyme is provided in an inactive state and upon thermal activation, the modifier is permanently released, regenerating active enzyme. The resulting hot-start PCR amplification provides increased sensitivity, specificity, and yield over conventional PCR techniques.

AmpliTaq Gold DNA Polymerase can be activated partially or completely in a pre-PCR heat step or can be allowed to activate slowly in a time-released manner during the denaturation steps of thermal cycling. With or without a limited up-front heat activation step, active enzyme is released slowly during thermal cycling to match template concentration and increase specificity. The yield of specific product increases as reactants are not wasted in the formation of unintended products.

Features

  • Automated chemical hot-start enzyme for increased specificity, sensitivity, and yield
  • Time-released thermal activation improves sensitivity in low copy number amplifications
  • Successful multiplex PCR saves time and reagents
  • Includes GeneAmp 10X PCR Buffer II with MgCl2
  • Hot-start technology reduces risk of biological contamination

Notes

  • For better performance AmpliTaq Gold 360 DNA Polymerase is purified by an additional proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation.
  • When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates.

Order Info

Shipping condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Content And Storage • AmpliTaq Gold DNA Polymerase (5 U/μL), 1 x 50 μL
• 10X PCR Buffer II, 1 x 1.5 mL
• 25 mM MgCl2, 1 x 1.5 mL

Store at -15°C to -30°C.
Format Tube
GC-Rich PCR Performance Low
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Standard
Exonuclease Activity 5' - 3'
Product Type DNA Polymerase
Quantity 250 Units
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 1X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang 3'-A
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.