missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Amplitude™ Optic Wipes
Optic wipes made of 100% nonwoven cellulose with long staple fibers, non-abrasive surface texture, low levels of extractables and only moderate particle generation, absorbent and optical grade, designed for delicate cleaning and lining of surfaces and trays
Brand: Contec™ C6-99
510.99 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Recommended for ISO Class 6-8 environments
- Long staple fibers create nonabrasive, absorbent wipe
- Nonwoven wipe with low levels of extractables
- Only moderate particle generation
- Ideal for use in controlled environments
- Specifically designed for delicate cleaning of optical products and microminiature electronic components
Specifications
Wipe | |
23 cm | |
23 cm | |
Process tools and other equipment |
9 in. | |
9 in. | |
Cellulose | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction