missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ampule Breakers

Thermo Scientific Ampule Breakers provide a safe, convenient way to open 1 mL to 10 mL glass ampules.

Brand:  Thermo Scientific™ 28916

 View more versions of this product

Product Code. 16365212

  • 37.15 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These single-use polyethylene sleeve ampule breakers help protect your fingers from glass shards when opening 1 mL to 10 mL glass ampules. The Thermo Scientific Ampule Breakers encapsulate the ampule top upon opening, making disposal of the glass top easy and safe.

• Easy to use single-use format
• Compatible with 1 mL to 10 mL glass ampules
• Protects against glass shards
• Helps prevent cross contamination

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
Ampule Breaker
5-10 mL
100 Breakers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.