Learn More
Agilent BioTek AMX Cassette
Two cassettes are required for use with the AMX module (PN 1260041). User installs cassettes with AMX manifolds (included with AMX module). For use with MFXP2R, order two cassettes (does not require PN 1260041).
Brand: Agilent BioTek 1260035
This item is not returnable.
View return policy
Description
Dispensing speeds:
- 96 wells, 5 μL cass, 10 μL/ well: 3 seconds
- 96 wells, 5 μL cass, 20 μL/ well: 3.5 seconds
- 384 wells, 5 μL cass, 5 μL/ well: 6.5 seconds
- 384 wells, 10 μL cass, 10 μL/ well: 8 seconds
- 384 wells, 1 μL cass, 1 μL/ well: 6 seconds
- 1536 wells, 1 μL cass, 1 μL/ well: 21 seconds
Specifications
<5% | |
2 D and 3 D Cell Culture to Concentration Normalization Assays, ELISA, Bead-Based Assays and More | |
Two 8-channel autoclavable manifolds, tube-in-tube design | |
50/60 Hz |
Autoclavable | |
MFPX2 | |
<5% CV |