missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amyloid beta 40 Human ELISA Kit

Quantifies natural and recombinant human Aβ40 (Beta Amyloid 40) selectively in tissue culture medium, tissue homogenate, and cerebrospinal fluid (CSF)

Brand:  Invitrogen™ KHB3482

 View more versions of this product

Product Code. 10681484

  • 1456.00 € / 192 tests
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Human Amyloid Beta 40 (Hu Abeta 40) ELISA quantitates Hu Abeta 40 in human tissue culture medium, tissue homogenate, and cerebrospinal fluid (CSF). The assay will exclusively recognize both natural and recombinant Hu Abeta 40. Principle of the method The Human Abeta 40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Beta-amyloid is a 40–43 amino acid peptide cleaved from amyloid precursor protein, and a putative gamma-secretase. Amyloid beta (1-40) together with Amyloid beta (1-42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the amyloid beta (40) and amyloid beta (42) alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.

Order Info

Shipping condition: Wet ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

P05067
<6 pg/mL
Biotin
Cerebrospinal Fluid, Supernatant, Tissue Homogenates
Colorimetric Microplate Reader
AAA,ABETA,ABPP,AD1I,CTFgamma,CVAP,PN-II,PN2,APP
3.8%
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Aβ 40
RUO
2°C to 8°C
1 hr. 20 min.
7.8-500 pg/mL
9.8 to 500pg/mL
ELISA Kit
Human
351
4 hr.
2.5%
HRP
2 x 96 Tests
Cerebrospinal Fluid,50 μL; Supernatant, 50 μL; Tissue Homogenate,50 μL
Human
4 hr.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.