Learn More
BINDER™ Analog Output
Non-adjustable analog output
Brand:  BINDER™ 8012-2573
 View more versions of this product
Product Code. 30238254
  • 729.00 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Analog output
  • Not adjustable
  • For temperature values or temperature and humidity values
  • 4 to 20 mA
Specifications
Specifications
KB PRO 130, KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470
Copper
Analog Output
For Temperature Values
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Analog Output > For use with KB PRO 130, KB PRO 260, KB PRO 470, KB PRO 260, KB PRO 470