Learn More
OHAUS™ Analytical Balance - PR Series
Designed for Routine Weighing Applications in Your Workplace
Brand:  OHAUS™ 30524385
1519.83 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 16650324
  • 1685.00 € / Each
Explore more special offers
Description
Description
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
InCal™ - Semi Automatic
RS232 (one port included)
Backlit LCD
80% RH
PR224M
3.000 sec.
317 x 201 x 303 mm
317.00 mm
201.00 mm
303.000 mm
317.00 mm
4.500 kg
User-selectable span calibration points
User-selectable environmental filters and brightness settings, auto-tare, auto-dim, user-selectable span calibration points, software lockout and reset menu, user-selectable communication settings and data print options, user-definable project and user IDs, software overload/underload indicator, stability indicator
0.0001 g
0.0002 g
0.0001 g
Milligram, Gram, Carat
12.50 in.
7.90 in.
11.90 in.
12.50 in.
10.000 lb.
AC Adapter (Included)