Learn More
Abeba™ Anatom 32120 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32120/49
 View more versions of this product
Product Code. 15495804
  • 112.00 € / 1 pair
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • suitable for heel and foot lift
  • Suitable for hygienically demanding areas
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E SRC
Microfiber
Slip-resistant
32120
White
49
Polyurethane
Steel