Learn More
Abeba™ Anatom 32125 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
112.00 €
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 SB A, E SRC |
---|---|
Color | Black |
Material | Microfiber |
Sole Features | Slip-resistant |
Sole Material | Polyurethane |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15649974
|
Abeba™
32125/36 |
36 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15659974
|
Abeba™
32125/37 |
37 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15669974
|
Abeba™
32125/38 |
38 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15679974
|
Abeba™
32125/39 |
39 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15689974
|
Abeba™
32125/40 |
40 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15699974
|
Abeba™
32125/41 |
41 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15609984
|
Abeba™
32125/42 |
42 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15619984
|
Abeba™
32125/43 |
43 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15629984
|
Abeba™
32125/44 |
44 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15639984
|
Abeba™
32125/45 |
45 |
112.00 €
1 pair |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
15649984
|
Abeba™
32125/46 |
46 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15659984
|
Abeba™
32125/47 |
47 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15669984
|
Abeba™
32125/48 |
48 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15679984
|
Abeba™
32125/49 |
49 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15689984
|
Abeba™
32125/50 |
50 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15699984
|
Abeba™
32125/51 |
51 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
15609994
|
Abeba™
32125/52 |
52 |
112.00 €
1 pair |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20345:2011 SB A, E SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3582)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- Suitable for heel and foot lift
- Suitable for hygienically demanding areas
Specifications
CE EN ISO 20345:2011 SB A, E SRC | |
Microfiber | |
Polyurethane | |
Steel |
Black | |
Slip-resistant | |
32125 |