Learn More
Abeba™ Anatom 32130 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32130/52
120.40 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.
 View more versions of this product
Product Code. 15617678
  • 121.00 € / 1 pair
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber
  • White
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • velcro fastening
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
32130
White
52
Polyurethane
Steel