Learn More
ABEBA™ Anatom Black/Red Safety Shoes S3 ESD
Red/black Anatom shoes made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Brand: ABEBA™ 32238/40
This item is not returnable.
View return policy
Description
- Comfortable, robust and durable shoes
- Comply to EN ISO 20345: 2012, S3, SRC
- ESD dissipative according to EN ISO 61340
- Slip resistant PU double density outsole
- Black/red water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifications
Black, Red | |
40 | |
EN ISO 20345: 2012 S3 SRC, EN ISO 61340 ESD class 1 | |
Low-cut | |
S3 SRC ESD |
Cordura | |
Steel | |
Men's | |
Water Resistant |