Learn More
IKA™ R 1333 Anchor Stirrer
Overhead, anchor-shaped stirrer element is ideally suited for medium to highly viscous fluids.
Brand:  IKA™ 0002747400
Product Code. 10186810
  • 913.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tangential flow
  • High shearing rate at edges
  • Minimum deposits on the vessel wall
  • Use at low speeds
  • Use for polymer reactions, even distribution of high mineral contents in liquids
  • Ideal for medium to highly viscous fluids
Specifications
Specifications
R 1333
0.4 in.
10 mm
550 mm
Anchor Impeller
5.9 in.
150 mm
21.7 in.
800 rpm