missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Angled Two-Neck Round-Bottom Flask

Distillation flasks with angled side neck (20°)

Brand:  Lenz Laborglasintrumente™ 03214270

 View more versions of this product

Product Code. 10284571

  • 74.00 € / Each
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Distillation flasks acc. to DIN 12394 and in addition to DIN standard
  • With NS
  • With angled side neck (20°)
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

Center neck - NS 29/32, Side Neck - NS 19/26
Borosilicate Glass (3.3)
131 mm
1000 mL
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.