missing translation for 'onlineSavingsMsg'
Learn More
Anhydrotetracycline, IBA Lifesciences Product Code.: 12813196
compliance-icons compliance-icons

Anhydrotetracycline, IBA Lifesciences
GREENER_CHOICE

Product Code. 12813196 Shop All IBA Lifesciences Products
Quantity: 25mg
Click to view available options
Quantity: 25mg
This item is not returnable. View return policy

Product Code. 12813196

Brand: IBA Lifesciences 20401002

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Derivative of tetracycline for induction of tetracycline-controlled promoters

Anhydrotetracycline is a derivative of tetracycline and an efficient inducer of the tetracycline promoter present in E. coli expression vectors pASK-IBA and pASG-IBA. It does not exhibit antibiotic activity within our recommended concentrations. 
Greener Choice Claims
  • Environmental benefits include:
    • Non-Toxic

Specifications

CAS 1665-56-1
Color Yellow
Molecular Formula C22H22N2O7
Quantity 25 mg
Synonym anhydrotetracycline, 5 A,6-anhydrotetracycline, 4 s-anhydrotetracycline, cxcveerymjzmmm-docrcchosa-n, 4 s,4 AS,12 AS-4-dimethylamino-3,10,11,12 A-tetrahydroxy-6-methyl-1,12-dioxo-1,4,4 A,5,12,12 A-hexahydrotetracene-2-carboxamide
Solubility Information aq. soln. 1:200 m/v
InChI Key JGRMImmCZBOWCW-LCGGBVFXSA-N
Molecular Weight (g/mol) 426.43
PubChem CID 54675758
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Anhydrotetracycline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anhydrotetracycline, IBA Lifesciences >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.