missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aniline Blue, sodium salt, pure, water soluble, certified

Product Code. 10556592
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 28983-56-4
Absorption Ratio 0.96 to 1.09
Concentration 12.0mL of 0.1N TiCl3/g min.
Molecular Formula C37H26N3Na2O9S3
MDL Number MFCD00058509
Synonym Acid Blue 22, C.I. 42755
InChI Key TUHAIJABPUJAEY-UHFFFAOYSA-K
Infrared Spectrum Authentic
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O
IUPAC Name disodium 2-{[4-({4-[(2-sulfonatophenyl)amino]phenyl}({4-[(2-sulfonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene})methyl)phenyl]amino}benzene-1-sulfonate
Molecular Weight (g/mol) 798.79
PubChem CID 131675892
Lambda Max 594 to 610nm
Formula Weight 737.71
Percent Purity 12.0 mL of 0.1 N TiCl3/g min.
Grade Pure
Packaging Glass bottle
Vapor Pressure Vapor Pressure: Negligible
Color Brown/Purple
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Aniline Blue, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Aniline Blue, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : C.I. Acid blue 93

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'