Learn More
Thermo Scientific Chemicals Aniline Blue, sodium salt, pure, water soluble, high purity biological stain
64.70 € - 189.00 €
Chemical Identifiers
CAS | 28983-56-4 |
---|---|
Molecular Formula | C37H26N3Na2O9S3 |
Molecular Weight (g/mol) | 798.79 |
MDL Number | MFCD00058509 |
InChI Key | TUHAIJABPUJAEY-UHFFFAOYSA-K |
Synonym | Acid Blue 22, C.I. 42755 |
PubChem CID | 76956083 |
IUPAC Name | disodium;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfonatophenyl)azaniumylidenecyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonate |
SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10656822
|
Thermo Scientific Chemicals
229660250 |
25 g | Glass bottle |
64.70 €
25g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
10226570
|
Thermo Scientific Chemicals
229661000 |
100 g | Glass bottle |
189.00 €
100g |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
28983-56-4 | |
798.79 | |
TUHAIJABPUJAEY-UHFFFAOYSA-K | |
76956083 | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O |
C37H26N3Na2O9S3 | |
MFCD00058509 | |
Acid Blue 22, C.I. 42755 | |
disodium;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfonatophenyl)azaniumylidenecyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonate |
Specifications
28983-56-4 | |
C37H26N3Na2O9S3 | |
Acid Blue 22, C.I. 42755 | |
TUHAIJABPUJAEY-UHFFFAOYSA-K | |
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NC1=CC=C(C=C1)C(C1=CC=C(NC2=CC=CC=C2S([O-])(=O)=O)C=C1)=C1C=CC(C=C1)=NC1=CC=CC=C1S([O-])(=O)=O | |
798.79 | |
594 to 610nm (in pH 3 water) | |
Pure | |
Crystalline Powder |
0.96 to 1.09 | |
MFCD00058509 | |
Solubility in water: soluble. | |
Authentic | |
disodium;4-[4-[[4-(4-sulfoanilino)phenyl]-[4-(4-sulfonatophenyl)azaniumylidenecyclohexa-2,5-dien-1-ylidene]methyl]anilino]benzenesulfonate | |
76956083 | |
799.81 | |
Brown/Purple | |
Aniline Blue, sodium salt |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 249-113-9
missing translation for 'rtecsNumber' : DB4958000
missing translation for 'tsca' : TSCA
RUO – Research Use Only