Learn More
Masterflex™ Animal Free Luer Fitting, ADCF female luer to 1/8 in. L barb adapter
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Cole-Parmer™ Animal Free Luer Fittings offer superior chemical resistance and are suited for ETO and gamma sterilization.
Brand:  Masterflex™ 30800-08
Product Code. 13515770
  • 44.55 € / Pack of 25
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • ADCF Female Luer to 1/8 in. L Barb Adapter
  • Not recommended for autoclaving
  • Sold in packs of 25
Specifications
Specifications
Class VI
1/8 in.
Clear
Polypropylene
25/Pk.