missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Anios DTS™ Spray Bottles
ANIOS DTS cleans and removes stains from any washable surface. Removes: Ink stains, felt-tip pen, marker pen, pencil, ball-point pen, paint, lipstick, rubber, tannins, stubborn stains on lab benches, Instrubacs, etc.
Brand: Laboratoires Anios™ 2319512UG
This item is not returnable.
View return policy
Description
Multipurpose Stain Remover:
- Clear and colourless solution
- pH of the pure product at +20°C: approx. 10.6
- Density at + 20°C: approx. 1
Specifications
Anionic surfactants, alkaline agent, glycol derivatives, synthetic fragrance | |
Bottle | |
12/Pk. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ANIOS DTS
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : DOCUSATE SODIUM, MONOPROPYLENE GLYCOL METHYL ETHER, 2-AMINOETHANOL