missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Aniosgel 800 Sanitizer
Laboratoires Anios™ Aniosgel 800 Sanitizer is suitable for hygienic treatment and surgical disinfection of hands by rubbing.
72.41 € - 209.00 €
Specifications
Product Type | Sanitizer Gel |
---|---|
Certifications/Compliance (Europe) | EN 13727, EN 13727, EN14348, EN 13624, EN 14476 |
Composition | Ethanol in the presence of thickening, moisturizing and emollient agents, and water. |
For Use With (Application) | Hygienic treatment and surgical disinfection of hands by rubbing. |
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
15551823
|
Laboratoires Anios™
2550762GP |
Bottles with screw pump |
72.41 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15561823
|
Laboratoires Anios™
2550763GP |
Botte with Screw Pump |
209.00 €
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- High level of observance
- Tested under dermatological control
- Adapted to sensitive skin
- Preserves skin hydration
- Broad antimicrobial spectrum from 30 sec.
- Conforms to latest European standards
- Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Sanitizer Gel | |
EN 13727, EN 13727, EN14348, EN 13624, EN 14476 | |
Ethanol in the presence of thickening, moisturizing and emollient agents, and water. | |
Hygienic treatment and surgical disinfection of hands by rubbing. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- aniosgel 800
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
- P501-Dispose of contents/container to as hazardous waste.
Supplemental information
- MIXTURE LIST-Contains : ethyl alcohol, propan-2-ol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Laboratoires Anios™ Aniosgel 800 Sanitizer