missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosgel 85 Blue

Laboratoires Anios™ Aniosgel 85 Blue is a hydroalcoholic gel for hygienic treatment and disinfection of hands.

60.75 € - 127.00 €

Specifications

Product Type Sterilizer/Disinfectant Hand Gel
Certifications/Compliance (Europe) EN 1040, pr EN 12054, EN 1500, EN 12791, EN 14348, EN 1275, EN 1650, EN 14476+A1
Composition Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
For Use With (Application) Hygienic treatment and surgical disinfection of hands
Packaging Airless bottles
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15431121
View Documents
Laboratoires Anios™
1645274
75 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15441121
View Documents
Laboratoires Anios™
1645363
500 mL
127.00 €
Pack of 12
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
Description

Description

  • Broad antimicrobial spectrum
  • Hygienic treatment in 30 sec.
  • For hygienic treatment and surgical disinfection
  • Active against bacteria, tuberculosis, fungus, viruses
Specifications

Specifications

Sterilizer/Disinfectant Hand Gel
Ethanol in the presence of thickening, moisturizing and emollient agents, and water.
Airless bottles
EN 1040, pr EN 12054, EN 1500, EN 12791, EN 14348, EN 1275, EN 1650, EN 14476+A1
Hygienic treatment and surgical disinfection of hands
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol, propan-2-ol

SDS
Documents

Documents

Product Certifications