missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant
Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant is an ethanol-based, broad antimicrobial spectrum spray for rapid disinfection of surfaces and non-immersible, non-invasive equipment.
213.00 € - 217.00 €
Specifications
Product Type | Rapid Disinfectant |
---|---|
Certifications/Compliance | Bacteria: EN 13727, EN 1276, EN 13697, MRB according to EN 13697Mycobacteria: EN 14348, EN 14563Yeasts/Molds: EN 13624, EN 1650, EN 13697, EN 14562 : Tricophyton mentagrophytesViruses: Adenovirus, HIV-1, PRV (surrogate of HBV), BVDV (surrogate of HCV), Vaccinia virus Herpes-virus, Coronavirus, Rotavirus, Infl uenza virus A (H N ), Feline Calicivirus, Norovirus MNV according to EN 14476 |
Composition | Hydroalcoholic solution (ethanol 55%), quaternary ammonium propionate,fragrance. |
For Use With (Application) | Rapid disinfection of non-immersible medical devices resistant to alcohol. |
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Price | Quantity & Availability | ||||
15451884
|
Laboratoires Anios™
2084073 |
1 L | 12/Cs. |
217.00 €
Pack of 12 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15461884
|
Laboratoires Anios™
2084034 |
5 L | 4/Cs. |
213.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ethanol-based composition
- Broad activity spectrum
- Leaves no trace when drying
- Pleasant fragrance
- Aldehyde free, without coloring
- Rapid action disinfection of surfaces and previously cleaned, non-immersible medical devices resistant to alcohol (stethoscopes, pressure sensors, blood sugar testers, etc.)
- Active against bacteria, mycobacteria, yeasts, molds, viruses
Specifications
Rapid Disinfectant | |
Bacteria: EN 13727, EN 1276, EN 13697, MRB according to EN 13697Mycobacteria: EN 14348, EN 14563Yeasts/Molds: EN 13624, EN 1650, EN 13697, EN 14562 : Tricophyton mentagrophytesViruses: Adenovirus, HIV-1, PRV (surrogate of HBV), BVDV (surrogate of HCV), Vaccinia virus Herpes-virus, Coronavirus, Rotavirus, Infl uenza virus A (H N ), Feline Calicivirus, Norovirus MNV according to EN 14476 | |
Hydroalcoholic solution (ethanol 55%), quaternary ammonium propionate,fragrance. | |
Rapid disinfection of non-immersible medical devices resistant to alcohol. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ANIOSPRAY QUICK
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, PROPAN-2-OL, DIDECYLDIMETHYLAMMONIUM CHLORIDE
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Laboratoires Anios™ Aniospray Quick Spray Surface Disinfectant