Learn More
Anisil, 98+%, Thermo Scientific Chemicals
34.17 € - 375.87 €
Chemical Identifiers
CAS | 1226-42-2 |
---|---|
Molecular Formula | C16H14O4 |
Molecular Weight (g/mol) | 270.284 |
MDL Number | MFCD00008405 |
InChI Key | YNANGXWUZWWFKX-UHFFFAOYSA-N |
Synonym | 4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione |
PubChem CID | 71043 |
IUPAC Name | 1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
SMILES | COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453264
|
Thermo Scientific Alfa Aesar
A10594.06 |
5 g |
34.17 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463264
|
Thermo Scientific Alfa Aesar
A10594.14 |
25 g |
116.58 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11473264
|
Thermo Scientific Alfa Aesar
A10594.22 |
100 g |
375.87 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAnisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
1226-42-2 | |
270.284 | |
YNANGXWUZWWFKX-UHFFFAOYSA-N | |
71043 | |
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC |
C16H14O4 | |
MFCD00008405 | |
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
1,2-bis(4-methoxyphenyl)ethane-1,2-dione |
Specifications
1226-42-2 | |
C16H14O4 | |
531345 | |
Insoluble in water. | |
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC | |
270.284 | |
270.29 | |
Anisil |
132°C to 134°C | |
MFCD00008405 | |
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione | |
YNANGXWUZWWFKX-UHFFFAOYSA-N | |
1,2-bis(4-methoxyphenyl)ethane-1,2-dione | |
71043 | |
≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 214-960-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only