missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Mouse anti-Human, Mouse, Clone: 1E1B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66344-1-IG-20UL
This item is not returnable.
View return policy
Description
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.Specifications
Annexin A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04083, P10107 | |
Anxa1 | |
Annexin A1 Fusion Protein Ag17273 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
1E1B7 | |
Unconjugated | |
ANXA1 | |
Annexin 1, annexin A1, Annexin I, ANX1, ANXA1, Calpactin 2, Calpactin II, Chromobindin 9, Lipocortin I, LPC1, p35 | |
Mouse | |
Protein G | |
RUO | |
16952, 301 | |
-20°C | |
Liquid |