missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin Binding Buffer (5X), for flow cytometry

Annexin Binding Buffer (5X), for flow cytometry

Brand:  Invitrogen™ V13246

78.38 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 12063805

  • 85.00 € / 50mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

This product is designed to facilitate the binding of annexin V to phosphatidylserine for use in apoptosis assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mL
Annexin Binding Buffer
Wet Ice
Flow Cytometry
7.4
Contains 1 bottle of annexin binding buffer (5x solution, 50 mL).

Store at 2–8°C.
50 mM HEPES, 700 mM NaCl, 12.5 mM CaCl2, pH 7.4
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.