missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Annexin V Conjugates for Apoptosis Detection
Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.
Brand: Molecular Probes™ A35110
This item is not returnable.
View return policy
Description
Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.
Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.
In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.
The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.
The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits
Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.
Specifications
Annexin V, APC conjugate (APC Annexin V) (replaces Annexin product- AnnexinV05) | |
Contains 1 vial of annexin V, APC conjugate. | |
Store in refrigerator (2°C to 8°C) and protect from light. | |
650/660 | |
Red | |
APC (Allophycocyanin) |
250 μL | |
Annexin V conjugate | |
633-637 | |
50 | |
Wet Ice | |
Flow Cytometer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction