missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Annexin V Conjugates for Apoptosis Detection

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.

Brand:  Molecular Probes™ A35108

 View more versions of this product

Product Code. 10267452

  • 427.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.

Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Annexin V, Alexa Fluor 555 conjugate
Contains 1 vial of annexin V, Alexa Fluor 555 conjugate.
Store in refrigerator (2°C to 8°C) and protect from light.
555/565
Orange
Alexa Fluor 555
500 μL
Annexin V conjugate
532, 561
100
Wet Ice
Flow Cytometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.