Learn More
SCHOTT AG Lighting and Imaging™ Fiber Optic Ring Light
Fiber Optic Ring Lights are available as annular or darkfield versions with specific advantages. For all Ring Lights we offer adapters for a variety of objective series.
Brand:  SCHOTT AG Lighting and Imaging™ 157406
 View more versions of this product
Product Code. 12826553
  • 1095.00 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Annular Ring Lights offer absolute shadow-free illumination.
  • Darkfield Ring Lights offer low-angle illumination for shiny, reflective surfaces.
  • Ring Light with eco-friendly Puravis fiber optics.
  • High white light transmission thanks to the use of SCHOTT PURAVIS™ fibers.
  • Fiber bundle protected by flexible metal PVC sheating.
  • Mountable direct to the microscope objective.
  • Adapters for different objective diameters available, need to be ordered separately.
  • Housing made of rugged, black anodized aluminum.
Specifications
Specifications
Fiber Optic Ringlight
120 mm
Illumination for Stero-Microscopy. Offers low angle illumination for shiny, reflective surfaces.
1.4 in.
3.3 ft.
Eco-friendly Puravis Glass Optical Fibers
4.7 in.
66 to 70 mm
Cold Light Source KL1600LED, KL2500LED or KL1500HAL
36 mm
1000 mm for the flexible part
5 to 15 mm