missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
Brand: Ansell 2100100
152.53 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.
- Provide flexibility and tactility
- Made from an exceptionally thin chemical-resistant, five-layer laminated film
- Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
- Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
- Hand specific
- Broad spectrum chemical resistance
- Wing-thumb for added wearer comfort
Specifications
CE 0493, EN420:2003 + A1:2009; Category lll | |
Specific | |
Laminated Film | |
2.5 mil | |
EN 388:0000 | |
ABCEFGHIMS | |
Gauntlet | |
Unlined | |
AlphaTec™ 02-100 | |
38 cm |
White | |
Yes | |
10 | |
0.06 mm | |
Type A | |
Bacteria, Fungi WITH Virus Protection | |
Smooth | |
2-100 10 902213 | |
15 in. |