missing translation for 'onlineSavingsMsg'
Learn More

Ansell Hynit™ 32-125 Nitrile Coated Liquid Repellent Gloves

Product Code. 12650832
Click to view available options
Size:
7
7.5
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Special nitrile-coating formulation is flexible while also repelling liquid, oil, and dirt. Gloves are compliant with FDA Food Handling Requirements.

  • Liquid-repellent impregnation repels oil, grease, and dirt
  • Nitrile coating is flexible for optimum dexterity
  • Compliant with FDA food handling requirements
  • Resistant to abrasions
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Product Type Dip-coated Gloves
Size 8
For Use With (Application) Automotive Assembly, Equipment Operation, Metal Fabrication, and Mining
Glove Cuff Style Open
Liner Material Cotton/Polyester Blend
Packaging Type Polybag
Series 32-125
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.