missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 28-359 Nitrile Coated Heavyweight Gloves with NitraGold™ Cuff

Product Code. 12660812
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves combine the strength of aramid fibers with the grip and flexibility of Ansell's advanced nitrile coating technology. NitraGold&Trade; cuff provides mechanical protection against cuts and abrasion.

  • Heavyweight design provides a substantial level of protection
  • Kevlar™ aramid fiber construction is strong and durable
  • Nitrile coating is oil resistant and enhances grip capabilities
  • NitraGold™ cuff provides mechanical protection against cuts and abrasion
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Cuff Material NitraGold™
Coating Material Nitrile
Color Black, Gold
Cut Resistance Rating ANSI A3
Hand Specific
Product Type Dip-coated Gloves
Size 8
EN388 Standard 4321C
For Use With (Application) Automotive Assembly/Repair
Liner Material Aramid Fiber
Series ActivArmr™ 28-359
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'