missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves

Product Code. 12832253
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.

  • Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
  • Neoprene foam coating enhances grip in dry, wet, or oily conditions
  • Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
  • Ergonomic design is comfortable and flexible
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Arc Flash Level 2
Certifications/Compliance NFPA 70E Hazard Risk Category 2
Certifications/Compliance (Europe) EN 407: 412110
Coating Material Neoprene
Color Green, Black
Cut Resistance Rating ANSI A4
Hand Specific
Material Aramid Fiber
Size 10
EN388 Standard 2X42C
For Use With (Application) Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work
Glove Cuff Style Knit
Series ActivArmr™ 80-813
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.