missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ Solvex™ 37-676 Nitrile Chemical Resistant Gloves

Product Code. p-304920
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12660862

Brand: Ansell 37676070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nitrile gloves deliver optimal results in wet or dry work environments where chemical resistance is crucial.

  • High-performance nitrile compound
  • Protects against abrasions, cuts, snags and punctures
  • Does not contain protein allergens
  • Raised-diamond pattern for a secure grip on wet or slippery materials
  • Form-fitting fingers, contoured palm, and a tapered wrist create a comfortable, ergonomic fit
  • Flock-lining keeps hands cool and dry
  • Ideal for work in meat processing, parts degreasing, machining operations, wiping down parts and machinery, and handling hoses, belts and steel chains
  • Length: 13 in., Thickness: 15 mil
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) EN 1149
Chlorinated Yes
Applications Industrial
Color Green
Finger Thickness (English) 0.0169 in.
Finger Thickness (Metric) 0.43 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0169 in.
Palm Thickness (Metric) 0.43 mm
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:4101X, EN 388:4101
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLOPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Glove Cuff Style Gauntlet
Grip Design Reversed Lozenge
Liner Material Cotton Flock
Lining Lined
Model 37-676 7 117721
Series AlphaTec™ Solvex™ 37-676
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.