missing translation for 'onlineSavingsMsg'
Learn More
Anthracene, 99%, Thermo Scientific Chemicals Product Code.: 11449522

Anthracene, 99%, Thermo Scientific Chemicals

Product Code. 11449522
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11449522

Brand: Thermo Scientific Alfa Aesar A10203.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anthracene is used in the production of the red dyealizarin and other dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used in the production of the red dyealizarin and other dyes.

Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 214°C to 218°C
Density 1.25
Boiling Point 340°C to 342°C
Flash Point 121°C (249°F)
Quantity 100 g
UN Number UN3077
Beilstein 1905429
Merck Index 14,682
Solubility Information Soluble in alcohol,methanol (1.8g/100 g),hexane (0.37g/100 g). Insoluble in water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Anthracene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241907 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberA10203.22,A10203.14,A10203.36
Lot Number10238880 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10203.22,A10203.14,A10203.36
Lot Number10179388 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10203.22,A10203.14,A10203.36
Lot Number10159856 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10203.22,A10203.14,A10203.36
Lot Number10190202 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10203.22,A10203.14,A10203.36
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Anthracene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.