missing translation for 'onlineSavingsMsg'
Learn More

Anthracene, 99%

Product Code. 11944451 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Plastic bottle
Unit Size:
100g
10kg
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 11944451

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-12-7 | C14H10 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-12-7
Molecular Formula C14H10
Molecular Weight (g/mol) 178.23
MDL Number MFCD00001240
InChI Key MWPLVEDNUUSJAV-UHFFFAOYSA-N
Synonym paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767
PubChem CID 8418
ChEBI CHEBI:35298
IUPAC Name anthracene
SMILES C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 215.0°C to 218.0°C
Color Yellow
Boiling Point 340.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 05,657
Fieser 12,290; 15,99; 16,13
Merck Index 15,674
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/67mL alcohol - 1g/70mL methanol, 1g/200mL ether - 1g/62mL benzene, 1g/85mL chloroform - 1g/31mL carbon disulfide, 1g/86mL ccl4 - 1g/125mL toluene
Formula Weight 178.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Anthracene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Anthracene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.