missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthrone, 95%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 152.0°C to 157.0°C |
Boiling Point | 721.0°C |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass Bottle |
Quantity | 100 g |
Beilstein | 07,473 |
Merck Index | 15,682 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hot naoh solution, conc. sulfuric acid, soluble in alcohol, benzene, aceton, soluble in most organic solvents without, fluorescence |
Formula Weight | 194.23 |
Percent Purity | 95% |
Show More |
Safety and Handling

Product Identifier
- Anthrone
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H361d-Suspected of damaging the unborn child.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Anthrone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Anthrone, 95%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction