Learn More
Description
Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 155°C to 158°C |
Quantity | 25 g |
Beilstein | 1910173 |
Sensitivity | Light sensitive |
Merck Index | 14,691 |
Solubility Information | Soluble in acetone,hot benzene,concentrated sulfuric acid,hot dilute alkali and most organic solvents. Insoluble in water. |
Formula Weight | 194.23 |
Grade | ACS |
Physical Form | Crystalline |
Chemical Name or Material | Anthrone |
Safety and Handling

- Anthrone
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 9(10H)-Anthracenone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.