missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-BCL2L1, RAF1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0283.1Set
Includes: BCL2L1 rabbit purified polyclonal antibody (20μg); RAF1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the BCL2L1 protein, and the other against the RAF1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
BCL2L1 | |
Monoclonal/Polyclonal | |
BCL2L1 | |
BCL2L1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. BCL2L1 rabbit purified polyclonal antibody (20 ug) 2. RAF1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
BCL-XL/S/BCL2L/BCLX/Bcl-X/DKFZp781P2092/bcl-xL/bcl-xS | |
Mouse, Rabbit | |
RUO | |
598/5894 | |
Human | |
BCL2L1 & RAF1 Protein Protein Interaction Antibody Pair |