missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD16; Clone: 3G8; BD
346.00 € - 725.00 €
Specifications
Antigen | CD16 |
---|---|
Clone | 3G8 |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15863089
|
BD Biosciences
563784 |
25 Tests |
346.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15873089
|
BD Biosciences
563785 |
100 Tests |
725.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human polymorphonuclear leukocytes
Flow cytometry
Specifications
CD16 | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Primary |
3G8 | |
Brilliant Ultraviolet 395 | |
RUO | |
Human polymorphonuclear leukocytes | |
Affinity Purified |